2-Methylnaphthalene 96%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: β-Methylnaphthalene
127170050 127175000 127170025
ACRO127170050EA 27.6 EUR
ACRO127170050 ACRO127175000 ACRO127170025
2-Methylnaphthalene 96%
2-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 241…242 °C (760 mmHg)
Melting Pt: 32…36 °C
Density: 1.01 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004118
CAS Number: 91-57-6
EINECS: 202-078-3
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≥95 %