2-Methylnaphthalene ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: β-Methylnaphthalene
L02459.22 L02459.36
L02459.22EA 25.1 EUR
L02459.22 L02459.36
2-Methylnaphthalene ≥97%
2-Methylnaphthalene
Purity: 97% CAS No.: 91-57-6 Molecular Formula: C11H10 Formula Weight: 142.20 Flash Point: 97°(206°F) Odour: Characteristic Boiling Point: 241-243° Beilstein Reference: 906859 UN No.: 3077 MDL No.: MFCD00004118
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 241…243 °C
Melting Pt: 31…35 °C
Density: 1.001
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004118
CAS Number: 91-57-6
EINECS: 202-078-3
UN: 3077
ADR: 9 III