Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Butylene oxide, THF, Diethylene oxide
Packaged under argon in resealable ChemSeal™ bottles.
41820.AK
41820.K2
41820.AKEA
49.1
EUR
41820.AK
41820.K2
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Tetrahydrofuran
Unstabilized
Purity: 99.8+%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Formula:
C₄H₈O Boiling Pt: 66 °C Melting Pt: –108 °C Density: 0.889 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II REACH: 05-2114615188-48 Merck Index: 14,9211 |