2-Bromopyridine
Supplier: Thermo Scientific
CC04010.10 CC04010.50
MAYBCC04010.10EA 0 EUR
MAYBCC04010.10 MAYBCC04010.50
2-Bromopyridine
2-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192
Melting Pt: –40.1 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
EINECS: 203-641-6
UN: 2810
ADR: 6.1 II