3,5-Dichloro-2,4,6-trifluoropyridine
Supplier: Thermo Scientific
Warning
Synonyms: 3,5-Dichlorotrifluoropyridine
SB01768.1 SB01768.10 SB01768.50
MAYBSB01768.1EA 0 EUR
MAYBSB01768.1 MAYBSB01768.10 MAYBSB01768.50
3,5-Dichloro-2,4,6-trifluoropyridine
3,5-Dichloro-2,4,6-trifluoropyridine
Formula: C₅Cl₂F₃N
MW: 201.96 g/mol
Boiling Pt: 159…160 °C (1013 hPa)
Melting Pt: 24…26 °C
Density: 1.62 g/cm³ (20 °C)
MDL Number: MFCD00006226
CAS Number: 1737-93-5
EINECS: 217-088-3
UN: 2811
ADR: 6.1,II