2-Chloro-3-nitropyridine

Supplier: Thermo Scientific
Warning

SB00655.1 SB00655.10 SB00655.50
MAYBSB00655.1EA 0 EUR
MAYBSB00655.1 MAYBSB00655.10 MAYBSB00655.50
2-Chloro-3-nitropyridine
2-Chloro-3-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR