2-Chloro-5-nitropyridine

Supplier: Thermo Scientific
Warning

SB00747.1 SB00747.10 SB00747.50
MAYBSB00747.1EA 0 EUR
MAYBSB00747.1 MAYBSB00747.10 MAYBSB00747.50
2-Chloro-5-nitropyridine
2-Chloro-5-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR