2-Chloro-3,5-dinitropyridine 98+%
Supplier: Thermo Fisher Scientific
Danger
208190010 208190050 208190250
ACRO208190010EA 33.5 EUR
ACRO208190010 ACRO208190050 ACRO208190250
2-Chloro-3,5-dinitropyridine 98+%
2-Chloro-3,5-dinitropyridine
Formula: C₅H₂ClN₃O₄
MW: 203.54 g/mol
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006233
CAS Number: 2578-45-2
EINECS: 219-937-3
UN: 2811
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 61 to 66 °C
GC ≥98.0 %