4-Aminopyridine

Supplier: Molekula

MOLE11822791-100GEA 130 EUR
MOLE11822791-100G
4-Aminopyridine
4-Aminopyridine
Formula: C₅H₆N₂
MW: 94,12 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 157…160 °C
Density: 1,25 g/cm³ (20 °C)
Flash Pt: 156 °C
MDL Number: MFCD00006439
UN: 2671
ADR: 6.1,II
Merck Index: 13,03964

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR