2-(Methylamino)pyridine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 4597-87-9
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 87°(188°F)
Refractive Index: 1.5780
Melting Point: 14-15°
Boiling Point: 200-201°
Beilstein Reference: 108188
UN No.: 2810
MDL No.: MFCD00006250
Formula:
C₆H₈N₂ Boiling Pt: 200…201 °C Melting Pt: 14…15 °C Density: 1.060 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006250 CAS Number: 4597-87-9 EINECS: 224-997-9 UN: 2810 ADR: 6.1,III |