4-Ethylpyridine 98%
Supplier: Thermo Scientific
Warning
150570250 150571000 150575000
ACRO150570250EA 33.8 EUR
ACRO150570250 ACRO150571000 ACRO150575000
4-Ethylpyridine 98%
4-Ethylpyridine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 168.3 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.942 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00006450
CAS Number: 536-75-4
EINECS: 208-646-7
UN: 1992
ADR: 3,III
Merck Index: 13,03880
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5009 to 1.5029 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %