4-Picoline 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Methylpyridine, γ-Picoline
157600010 157600025 157602500
ACRO157600010EA 58 EUR
ACRO157600010 ACRO157600025 ACRO157602500
4-Picoline 99%
4-Picoline
Formula: C₆H₇N
Boiling Pt: 145 °C (760 mmHg)
Melting Pt: 2.4 °C
Density: 0.95 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00006440
CAS Number: 108-89-4
EINECS: 203-626-4
UN: 2313
ADR: 3,III
Merck Index: 12,07556
Order Now
Specification Test Results
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %