2,6-Di-tert-butylpyridine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
219584-5G 219584-1G
SIAL219584-5GEA 167 EUR
SIAL219584-5G SIAL219584-1G
2,6-Di-tert-butylpyridine, Sigma-Aldrich®
2,6-Di-tert-butylpyridine
Formula: C₁₃H₂₁N
MW: 191.32 g/mol
Boiling Pt: 209 °C (1013 hPa)
Density: 0.852 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6
Merck Index: 12,03086