2-Phenylpyridine ≥98%
Supplier: Thermo Fisher Scientific
Warning
L07861.09 L07861.18
L07861.09EA 91.9 EUR
L07861.09 L07861.18
2-Phenylpyridine ≥98%
2-Phenylpyridine
Purity: 98% CAS No.: 1008-89-5 Molecular Formula: C11H9N Formula Weight: 155.20 Flash Point: 110°(230°F) Refractive Index: 1.6240 Melting Point: -5° Boiling Point: 268-270° Beilstein Reference: 110445 MDL No.: MFCD00006280
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 268…270 °C
Melting Pt: –5 °C
Density: 1.086
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
UN: 2810
ADR: 6.1 III