3-Phenylpyridine ≥97%
Supplier: Thermo Fisher Scientific
Warning
B23930.06 B23930.14
B23930.06EA 63.7 EUR
B23930.06 B23930.14
3-Phenylpyridine ≥97%
3-Phenylpyridine
Purity: 97% CAS No.: 1008-88-4 Molecular Formula: C11H9N Formula Weight: 155.20 Flash Point: 110°(230°F) Refractive Index: 1.6160 Boiling Point: 272° Beilstein Reference: 110400 MDL No.: MFCD00006380
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 272 °C
Melting Pt: 76…77 °C
Density: 1.085
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006380
CAS Number: 1008-88-4
EINECS: 213-762-6