2-Bromo-5-chlorothiophene ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
A16682.06
A16682.14
A16682.06EA
35
EUR
A16682.06
A16682.14
2-Bromo-5-chlorothiophene ≥97%
2-Bromo-5-chlorothiophene
Purity: 97%
CAS No.: 2873-18-9
Molecular Formula: C4H2BrClS
Formula Weight: 197.48
Flash Point: 85°(185°F)
Refractive Index: 1.5960
Boiling Point: 69-70°/18mm
Beilstein Reference: 109897
UN No.: 2810
MDL No.: MFCD00014523
Formula:
C₄H₂BrClS Boiling Pt: 69…70 °C (18 mmHg) Melting Pt: –2 °C Density: 1.803 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014523 CAS Number: 2873-18-9 EINECS: 220-708-5 UN: 2810 ADR: 6.1,III |