2-Chlorothiophene

Supplier: Molekula

MOLE23295014-100GEA 0 EUR
MOLE23295014-100G
2-Chlorothiophene
2-Chlorothiophene
Formula: C₄H₃ClS
MW: 118,59 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1,285 g/cm³ (20 °C)
MDL Number: MFCD00005421
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR