2-Methylthiophene

Supplier: Thermo Scientific
Danger

SB01500.1 SB01500.10 SB01500.50
MAYBSB01500.1EA 0 EUR
MAYBSB01500.1 MAYBSB01500.10 MAYBSB01500.50
2-Methylthiophene
2-Methylthiophene
Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 113 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR