2-Methylthiophene

Supplier: Molekula

MOLE22590573-100GEA 0 EUR
MOLE22590573-100G
2-Methylthiophene
2-Methylthiophene
Formula: C₅H₆S
MW: 98,17 g/mol
Boiling Pt: 113 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1,02 g/cm³ (20 °C)
MDL Number: MFCD00005451
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR