1-Bromoadamantane

Supplier: Thermo Scientific

Synonyms: 1-Adamantyl bromide

BTB13004.1 BTB13004.10 BTB13004.50
MAYBBTB13004.1EA 0 EUR
MAYBBTB13004.1 MAYBBTB13004.10 MAYBBTB13004.50
1-Bromoadamantane
1-Bromoadamantane
Formula: C₁₀H₁₅Br
MW: 215.13 g/mol
Boiling Pt: 226
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00074721
CAS Number: 768-90-1
EINECS: 212-200-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR