2-Bromoadamantane ≥98%
Supplier: Thermo Fisher Scientific
B22760.06
B22760.14
B22760.06EA
64.1
EUR
B22760.06
B22760.14
2-Bromoadamantane ≥98%
2-Bromoadamantane
Purity: 98%
CAS No.: 7314-85-4
Molecular Formula: C10H15Br
Formula Weight: 215.14
Melting Point: 136-141°
Beilstein Reference: 4655839
Formula:
C₁₀H₁₅Br Boiling Pt: 230 °C (1013 hPa) Melting Pt: 136…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074745 CAS Number: 7314-85-4 EINECS: 230-777-3 |