2-Adamantanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Adamantan-2-one
M-Clarity™ quality level = MQ100
146048-100G 146048-25G 146048-5G
SIAL146048-100GEA 174 EUR
SIAL146048-100G SIAL146048-25G SIAL146048-5G
2-Adamantanone, Sigma-Aldrich®
2-Adamantanone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 43 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2