1-Chloroadamantane 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
264240100
264240500
264242500
ACRO264240100EA
30.4
EUR
ACRO264240100
ACRO264240500
ACRO264242500
1-Chloroadamantane 98%
1-Chloroadamantane
Formula:
C₁₀H₁₅Cl Melting Pt: 163…166 °C Density: 1,26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00075627 CAS Number: 935-56-8 EINECS: 213-305-0 |
Specification Test Results
Appearance | White to light grey Adhering crystalline solid and/or chunks |
Infrared spectrum | Conforms |
GC | ≥97.5 % |