5,6-Epoxy-1-hexene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Epoxy-5-hexene
M-Clarity™ quality level = MQ100
260347-25G 260347-5G
SIAL260347-25GEA 215 EUR
SIAL260347-25G SIAL260347-5G
5,6-Epoxy-1-hexene, Sigma-Aldrich®
5,6-Epoxy-1-hexene
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 119…121 °C (1013 hPa)
Melting Pt: –156 °C
Density: 0.869 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010051
CAS Number: 10353-53-4
EINECS: 233-771-9
UN: 3271
ADR: 3,II