2-Pyrrolidone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pyrrolidin-2-one
P74370-1KG P74370-25G P74370-500G
SIALP74370-1KGEA 67.6 EUR
SIALP74370-1KG SIALP74370-25G SIALP74370-500G
2-Pyrrolidone, Sigma-Aldrich®
2-Pyrrolidone
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 245
Melting Pt: 24…26 °C
Density: 1.1115 g/cm³ (20 °C)
Flash Pt: 129
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 12,08200