Azetidine 98%
Supplier: Thermo Scientific
Danger
296730010 296732500 296730050
ACRO296730010EA 155 EUR
ACRO296730010 ACRO296732500 ACRO296730050
Azetidine 98%
Azetidine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 61…62 °C (760 mmHg)
Density: 0.85 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005165
CAS Number: 503-29-7
EINECS: 207-963-8
UN: 2733
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %