Palmitoleic acid 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(Z)-Hexadec-9-enoic acid
376910010
376910050
376912500
ACRO376910010EA
214
EUR
ACRO376910010
ACRO376910050
ACRO376912500
Palmitoleic acid 99%
Palmitoleic acid
Formula:
C₁₆H₃₀O₂ Boiling Pt: 162 °C (.6 mmHg) Melting Pt: .5 °C Density: 0.894 g/cm³ Flash Pt: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004437 CAS Number: 373-49-9 EINECS: 206-765-9 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4590 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...