Palmitoleic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: (Z)-Hexadec-9-enoic acid

M-Clarity™ quality level = MQ300

P9417-1G P9417-100MG P9417-5G
SIALP9417-1GEA 283 EUR
SIALP9417-1G SIALP9417-100MG SIALP9417-5G
Palmitoleic acid, Sigma-Aldrich®
Palmitoleic acid
Formula: C₁₆H₃₀O₂
MDL Number: MFCD00004437
CAS Number: 373-49-9
EINECS: 206-765-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR