Diisobutylaluminium hydride 1 mol/l in dichloromethane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
M-Clarity™ quality level = MQ200
214973-100ML 214973-1L 214973-800ML
SIAL214973-100MLEA 86 EUR
SIAL214973-100ML SIAL214973-1L SIAL214973-800ML
Diisobutylaluminium hydride 1 mol/l in dichloromethane, Sigma-Aldrich®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2,I