Diisobutylaluminium hydride 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
M-Clarity™ quality level = MQ200
214981-100ML 214981-800ML
SIAL214981-100MLEA 72 EUR
SIAL214981-100ML SIAL214981-800ML
Diisobutylaluminium hydride 1 mol/l in THF, Sigma-Aldrich®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: -80
Density: 0.798
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3 I