3,3'-Dithiodipropionic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,3-Dithiodipropionic Acid
109010-250G 109010-50G
SIAL109010-250GEA 84.8 EUR
SIAL109010-250G SIAL109010-50G
3,3'-Dithiodipropionic acid, Sigma-Aldrich®
3,3'-Dithiodipropionic acid
Formula: C₆H₁₀O₄S₂
MW: 210.28 g/mol
MDL Number: MFCD00002780
CAS Number: 1119-62-6
EINECS: 214-284-0