4-Nitrophenyl palmitate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Nitrophenyl hexadecanoate
M-Clarity™ quality level = MQ200
N2752-1G
N2752-5G
SIALN2752-1GEA
72.5
EUR
SIALN2752-1G
SIALN2752-5G
4-Nitrophenyl palmitate, Sigma-Aldrich®
4-Nitrophenyl palmitate
Formula:
C₂₂H₃₅NO₄ MW: 377.52 g/mol |
MDL Number:
MFCD00047732 CAS Number: 1492-30-4 EINECS: 216-084-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...