2-Imidazolidinethione 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione
122030010 122032500
ACRO122030010EA 127 EUR
ACRO122030010 ACRO122032500
2-Imidazolidinethione 98%
2-Imidazolidinethione
Formula: C₃H₆N₂S
MW: 102.16 g/mol
Melting Pt: 197…201 °C
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00005276
CAS Number: 96-45-7
EINECS: 202-506-9
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 196 to 200 °C
HPLC ≥97.5 %
Stabilizer Approx 500 ppm hydroquinone