2-Imidazolidinethione, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione
M-Clarity™ quality level = MQ200
03940-100G
03940-500G
SIAL03940-100GEA
120
EUR
SIAL03940-100G
SIAL03940-500G
2-Imidazolidinethione, Sigma-Aldrich®
2-Imidazolidinethione
Formula:
C₃H₆N₂S MW: 102.16 g/mol |
MDL Number:
MFCD00005276 CAS Number: 96-45-7 EINECS: 202-506-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...