2-Methoxybenzothiazole, Sigma-Aldrich®

Supplier: Merck

707104-1G 707104-5G
SIAL707104-1GEA 44 EUR
SIAL707104-1G SIAL707104-5G
2-Methoxybenzothiazole, Sigma-Aldrich®
2-Methoxybenzothiazole
Formula: C₈H₇NOS
MW: 165.22 g/mol
CAS Number: 63321-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR