Lithium aluminum hydride 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
M-Clarity™ quality level = MQ200
212776-100ML 212776-800ML 212776-4X25ML
SIAL212776-100MLEA 59.6 EUR
SIAL212776-100ML SIAL212776-800ML SIAL212776-4X25ML
Lithium aluminum hydride 1 mol/l in THF, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348