S-Ethyl thioacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W328200-100G W328200-1KG W328200-25G W328200-SAMPLE
SIALW328200-100GEA 238 EUR
SIALW328200-100G SIALW328200-1KG SIALW328200-25G SIALW328200-SAMPLE
S-Ethyl thioacetate, Sigma-Aldrich®
S-Ethyl thioacetate
Formula: C₄H₈OS
MW: 104.17 g/mol
MDL Number: MFCD00015178
CAS Number: 625-60-5
EINECS: 210-904-9