Cyclopentyl methyl ether (CPME), Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
675970-100ML 675970-1L 675970-2L
SIAL675970-100MLEA 70.9 EUR
SIAL675970-100ML SIAL675970-1L SIAL675970-2L
Cyclopentyl methyl ether (CPME), Sigma-Aldrich®
Cyclopentyl methyl ether (CPME)
Formula: C₆H₁₂O
Density: 0.86 g/cm³ (20 °C)
Flash Pt: –1 °C (closed cup)
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3,II