Guanidinium sulphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Guanidine sulphate

M-Clarity™ quality level = MQ100

307394-100G 307394-500G
SIAL307394-100GEA 81.2 EUR
SIAL307394-100G SIAL307394-500G
Guanidinium sulphate, Sigma-Aldrich®
Guanidinium sulphate
Formula: C₂H₁₂N₆O₄S
MW: 216.22 g/mol
MDL Number: MFCD00013115
CAS Number: 594-14-9
EINECS: 209-826-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR