9-Chloroacridine
Supplier: Thermo Scientific
KM08135.1
KM08135.10
KM08135.50
MAYBKM08135.1EA
0
EUR
MAYBKM08135.1
MAYBKM08135.10
MAYBKM08135.50
9-Chloroacridine
9-Chloroacridine
Formula:
C₁₃H₈ClN MW: 213.67 g/mol |
MDL Number:
MFCD00022266 CAS Number: 1207-69-8 EINECS: 214-895-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...