9-Chloroacridine, Sigma-Aldrich®

Supplier: Merck

159425-1G 159425-5G
SIAL159425-1GEA 55.4 EUR
SIAL159425-1G SIAL159425-5G
9-Chloroacridine, Sigma-Aldrich®
9-Chloroacridine
Formula: C₁₃H₈ClN
MW: 213.67 g/mol
MDL Number: MFCD00022266
CAS Number: 1207-69-8
EINECS: 214-895-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR