Acridine ≥97%
Supplier: Thermo Scientific
Warning
L01657.06 L01657.14
L01657.06EA 42.9 EUR
L01657.06 L01657.14
Acridine ≥97%
Acridine
Purity: 98+% CAS No.: 260-94-6 Molecular Formula: C13H9N Formula Weight: 179.22 Melting Point: 107-111° Boiling Point: 346° Merck Reference: 14,122 UN No.: 2713 MDL No.: MFCD00005025
Formula: C₁₃H₉N
MW: 179.22 g/mol
Boiling Pt: 346 °C
Melting Pt: 107…111 °C
Density: 1.005
Storage Temperature: Ambient
MDL Number: MFCD00005025
CAS Number: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: 6.1,III
Merck Index: 14,122