Hand protectors, Hot Grip
Supplier: Kartell
HotGrip
KART3915EA 59.4 EUR
KART3915
Hand protectors, Hot Grip
Hot Hand Protectors
These hand protectors made of silicone rubber provide protection against hot containers up to +260 °C and cold containers down to –57 °C. Internal relief surface.