Cadmium (II) chloride hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cadmium chloride hydrate
M-Clarity™ quality level = MQ100
529575-10G 529575-50G
SIAL529575-10GEA 69.7 EUR
SIAL529575-10G SIAL529575-50G
Cadmium (II) chloride hydrate, Sigma-Aldrich®
Cadmium (II) chloride hydrate
Formula: CdCl₂·H₂O
Melting Pt: 565 °C
Density: 3.327 g/cm³ (25 °C)
MDL Number: MFCD00149627
CAS Number: 35658-65-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,II