Divinylbenzene (mixture of isomers) stabilised, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Synonyms: DVB, Divinylbenzene
Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200
414565-250ML 414565-1L
SIAL414565-250MLEA 80.8 EUR
SIAL414565-250ML SIAL414565-1L
Divinylbenzene (mixture of isomers) stabilised, Technical Grade, Sigma-Aldrich®
Divinylbenzene (mixture of isomers)
Formula: C₁₀H₁₀
MW: 130.19 g/mol
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5