Methyl oxalyl chloride

Supplier: Molekula

MOLEM11177203EA 0 EUR
MOLEM11177203
Methyl oxalyl chloride
Methyl oxalyl chloride
Formula: C₃H₃ClO₃
MW: 122,51 g/mol
MDL Number: MFCD00000705
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR