2-Chloro-5-ethylpyrimidine, Sigma-Aldrich®

Supplier: Merck

457485-1G 457485-5G
SIAL457485-1GEA 22.9 EUR
SIAL457485-1G SIAL457485-5G
2-Chloro-5-ethylpyrimidine, Sigma-Aldrich®
2-Chloro-5-ethylpyrimidine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD00799503
CAS Number: 111196-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR