Ethyl mercaptoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ethyl 2-mercaptoacetate, Ethyl thioglycolate
M-Clarity™ quality level = MQ100
E34307-100G
E34307-25G
E34307-5G
SIALE34307-100GEA
112
EUR
SIALE34307-100G
SIALE34307-25G
SIALE34307-5G
Ethyl mercaptoacetate, Sigma-Aldrich®
Ethyl mercaptoacetate
Formula:
C₄H₈O₂S MW: 120.17 g/mol |
MDL Number:
MFCD00004874 CAS Number: 623-51-8 EINECS: 210-800-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...