2-Piperidinemethanol 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-(Hydroxymethyl)piperidine
168030250 168031000 168030050
ACRO168030250EA 145 EUR
ACRO168030250 ACRO168031000 ACRO168030050
2-Piperidinemethanol 97%
2-Piperidinemethanol
Formula: C₆H₁₃NO
Boiling Pt: 100…102 °C (8hPa)
Melting Pt: 58…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00005987
CAS Number: 3433-37-2
EINECS: 222-333-2
Order Now
Specification Test Results
Appearance White to light orange Solid
Infrared spectrum Conforms
GC ≥96.0 %