Chloromethyl chloroformate, Sigma-Aldrich®

Supplier: Merck

23181-10ML 23181-50ML
SIAL23181-10MLEA 195 EUR
SIAL23181-10ML SIAL23181-50ML
Chloromethyl chloroformate, Sigma-Aldrich®
Chloromethyl chloroformate
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
MDL Number: MFCD00077688
CAS Number: 22128-62-7
EINECS: 244-793-3
UN: 2745
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR